Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014122818.2 | XP_013978293.1 | 3 | 588 | 89 | |
XM_014122820.2 | XP_013978295.1 | 4 | 669 | 89 | ||
XM_014122821.2 | XP_013978296.1 | 4 | 704 | 89 | ||
XM_014122823.2 | XP_013978298.1 | 5 | 973 | 89 | ||
XM_022407690.1 | XP_022263398.1 | 2 | 983 | 89 | ||
XM_022407691.1 | XP_022263399.1 | 5 | 996 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |