Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632944.2 | XP_005633001.1 | 3 | 1573 | 137 | |
XM_005632945.3 | XP_005633002.1 | 3 | 1623 | 137 | ||
XM_542121.6 | XP_542121.2 | 3 | 1318 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |