Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXPH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625650.3 | XP_005625707.2 | 1046 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris |