Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314132.1 | NP_001301061.1 | 2 | 634 | 115 | |
GenBank mRNA | KF381338.1 | - | 2 | 634 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |