Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631912.3 | XP_005631969.1 | 3 | 895 | 91 | |
XM_005631913.2 | XP_005631970.1 | 3 | 737 | 91 | ||
XM_005631915.3 | XP_005631972.1 | 4 | 1083 | 91 | ||
XM_022406064.1 | XP_022261772.1 | 5 | 1233 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |