Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014106922.2 | XP_013962397.1 | 4 | 667 | 91 | |
XM_543001.4 | XP_543001.2 | 4 | 585 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |