Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534625.6 | XP_534625.2 | 4 | 2159 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |