Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534705.6 | XP_534705.2 | 2 | 276 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |