Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014117304.1 | XP_013972779.1 | 1 - 2 | 5686 | 141 | |
XM_022424348.1 | XP_022280056.1 | 1 - 2 | 5686 | 141 | ||
XM_022424349.1 | XP_022280057.1 | 1 - 2 | 5700 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons |