Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_844500.5 | XP_849593.1 | 2 - 3 | 743 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |