Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197169.1 | NP_001184098.1 | 3 | 256 | 162 | |
XM_005615765.3 | XP_005615822.1 | 4 | 416 | 162 | ||
GenBank mRNA | HM135452.1 | - | 3 | 256 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Amplicon spans exons and probe does not span exons |