Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048112.1 | NP_001041577.1 | 1 | 62 | 74 | |
XM_014107460.2 | XP_013962935.1 | 2 | 725 | 74 | ||
XM_022409564.1 | XP_022265272.1 | 2 | 726 | 74 | ||
XM_022409565.1 | XP_022265273.1 | 2 | 752 | 74 | ||
XM_022409566.1 | XP_022265274.1 | 2 | 639 | 74 | ||
XM_022409567.1 | XP_022265275.1 | 2 | 733 | 74 | ||
GenBank mRNA | DQ666280.1 | - | 1 | 62 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |