Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_541212.4 | XP_541212.1 | 6 | 2366 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |