Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204929.1 | NP_001191858.1 | 1 - 2 | 49 | 56 | |
XM_005617935.3 | XP_005617992.1 | 1 - 2 | 155 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |