Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM2AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204899.1 | NP_001191828.1 | 2 - 3 | 125 | 90 | |
XM_005626411.3 | XP_005626468.2 | 3 - 4 | 193 | 90 | ||
XM_005626413.3 | XP_005626470.2 | 3 - 4 | 193 | 90 | ||
XM_022424914.1 | XP_022280622.1 | 3 - 4 | 206 | 90 | ||
XM_022424915.1 | XP_022280623.1 | 3 - 4 | 202 | 90 | ||
XM_022424916.1 | XP_022280624.1 | 3 - 4 | 206 | 90 | ||
XM_022424917.1 | XP_022280625.1 | 3 - 4 | 193 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |