Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246744.1 | NP_001233673.1 | 4 - 5 | 335 | 69 | |
GenBank mRNA | JI886703.1 | - | 5 - 6 | 434 | 69 | |
JP038870.1 | - | 5 - 6 | 434 | 69 | ||
L40381.1 | - | 4 - 5 | 335 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |