Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246785.1 | NP_001233714.1 | 4 - 5 | 521 | 128 | |
GenBank mRNA | AY392425.1 | - | 4 - 5 | 521 | 128 | |
JI878574.1 | - | 1 | 553 | 128 | ||
JP043698.1 | - | 1 | 553 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |