Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513456.1 | XP_003513504.1 | 4 - 5 | 580 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Protease Detect Genomic DNA Probe spans exons |