Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lins | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499399.2 | XP_003499447.1 | 7 - 8 | 1591 | 68 | |
XM_007640152.1 | XP_007638342.1 | 7 - 8 | 1682 | 68 | ||
XM_007640153.1 | XP_007638343.1 | 7 - 8 | 1641 | 68 | ||
GenBank mRNA | JI882981.1 | - | 7 - 8 | 1613 | 68 | |
JP039043.1 | - | 7 - 8 | 1613 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |