Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641368.1 | XP_007639558.1 | 2 - 3 | 1179 | 72 | |
XM_007641369.1 | XP_007639559.1 | 3 - 4 | 3910 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |