Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502845.2 | XP_003502893.1 | 6 - 7 | 995 | 60 | |
GenBank mRNA | JI884520.1 | - | 1 - 2 | 54 | 60 | |
JP049700.1 | - | 1 - 2 | 54 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Phosphatase Probe spans exons |