Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650938.1 | XP_007649128.1 | 1 - 2 | 406 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |