Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495175.2 | XP_003495223.1 | 6 - 7 | 922 | 77 | |
GenBank mRNA | JI893171.1 | - | 3 - 4 | 235 | 77 | |
JP050547.1 | - | 3 - 4 | 235 | 77 |