Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100755320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512718.1 | XP_003512766.1 | 1 | 620 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |