Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504875.2 | XP_003504923.1 | 1 - 2 | 161 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |