Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn2aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641426.1 | XP_007639616.1 | 4 - 5 | 477 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |