Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripply2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495634.1 | XP_003495682.1 | 2 - 3 | 167 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Probe spans exons |