Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495942.2 | XP_003495990.1 | 2 - 3 | 1401 | 85 | |
XM_007645413.1 | XP_007643603.1 | 2 - 3 | 1233 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |