Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501795.2 | XP_003501843.1 | 1 - 2 | 811 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |