Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514536.1 | XP_003514584.1 | 2 - 3 | 200 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |