Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495127.2 | XP_003495175.1 | 4 - 5 | 457 | 65 | |
GenBank mRNA | JI877023.1 | - | 4 - 5 | 423 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |