Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515376.2 | XP_003515424.1 | 2 - 3 | 211 | 103 | |
XM_007607580.1 | XP_007605770.1 | 2 - 3 | 211 | 103 | ||
GenBank mRNA | JI887113.1 | - | 2 - 3 | 210 | 103 | |
JP051167.1 | - | 2 - 3 | 210 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |