Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifitm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650896.1 | XP_007649086.1 | 2 - 3 | 484 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |