Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501883.2 | XP_003501931.1 | 9 - 10 | 1222 | 75 | |
GenBank mRNA | JI892008.1 | - | 3 - 4 | 388 | 75 | |
JP055369.1 | - | 4 - 5 | 449 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |