Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496083.2 | XP_003496131.1 | 14 - 15 | 1922 | 83 |