Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495769.2 | XP_003495817.2 | 4 - 5 | 561 | 82 | |
XM_007643890.1 | XP_007642080.1 | 3 - 4 | 504 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Detect Genomic DNA Recommended Assay Probe spans exons |