Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509067.2 | XP_003509115.1 | 2 - 3 | 379 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |