Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496771.1 | XP_003496819.1 | 2 - 3 | 441 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |