Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502913.2 | XP_003502961.1 | 1 - 2 | 867 | 91 | |
XM_007643530.1 | XP_007641720.1 | 2 - 3 | 896 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |