Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leprel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646914.1 | XP_007645104.1 | 1 - 2 | 45 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |