Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499468.2 | XP_003499516.1 | 3 - 4 | 1410 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |