Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499952.2 | XP_003500000.1 | 1 - 2 | 115 | 92 | |
GenBank mRNA | JI872906.1 | - | 1 - 2 | 228 | 92 | |
JP047257.1 | - | 1 - 2 | 228 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |