Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515363.1 | XP_003515411.1 | 4 - 5 | 573 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |