Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654158.1 | XP_007652348.1 | 3 - 4 | 1122 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |