Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641080.1 | XP_007639270.1 | 4 - 5 | 427 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Probe spans exons |