Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509901.2 | XP_003509949.1 | 3 - 4 | 245 | 105 | |
XM_007650551.1 | XP_007648741.1 | 3 - 4 | 195 | 105 | ||
XM_007650552.1 | XP_007648742.1 | 4 - 5 | 308 | 105 | ||
XM_007650553.1 | XP_007648743.1 | 4 - 5 | 258 | 105 | ||
GenBank mRNA | JI874444.1 | - | 1 - 2 | 82 | 105 | |
JP050277.1 | - | 1 - 2 | 82 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |