Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cspg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644815.1 | XP_007643005.1 | 4 - 5 | 1606 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |