Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506659.1 | XP_003506707.1 | 3 - 4 | 357 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |