Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naaladl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642941.1 | XP_007641131.1 | 9 - 10 | 1403 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |