Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506487.2 | XP_003506535.1 | 3 - 4 | 1120 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |